Learn More
Aluminum magnesium isopropoxide, 99.95+% (metals basis)
CAS: 69207-83-6
Supplier: Thermo Scientific Chemicals 03932009
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Aluminum magnesium isopropoxide | |
| 40°C | |
| (metals basis) | |
| 10 g | |
| Moisture sensitive | |
| ≥99.95% |
| 69207-83-6 | |
| 114°C (0.2 mmHg) | |
| MFCD00070426 | |
| UN3181 | |
| 550.53 | |
| Crystalline Low Melting Solid |
Chemical Identifiers
| 69207-83-6 | |
| MFCD00070426 |
Safety and Handling
GHS H Statement
H225-H228-H315-H319-H335
Highly flammable liquid and vapor.
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only