missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum magnesium isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol
Description
Aluminum magnesium isopropoxide has been used to study the hydrolysis conditions on the properties of magnesium aluminate spinel powders.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 130°C to 142°C (2.0 mmHg) |
| Quantity | 25 mL |
| UN Number | UN1219 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Not miscible with water. |
| Formula Weight | 550.53 |
| Percent Purity | 99.9% |
| Physical Form | Liquid |
| Assay | (metals basis) |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H319-H335-H336 |
| Hazard Statement | GHS H Statement H225-H315-H319-H336 Highly flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only