missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Magnesium gauze , alloy 5056, 20 mesh woven from 0.23mm (0.009 in.) dia. wire, Thermo Scientific™

Catalog No. p-7080145
Change view
Click to view available options
Quantity:
150 x 150 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA46579GT 150 x 150 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA46579GT Supplier Thermo Scientific Chemicals Supplier No. 46579GT
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Name or Material Aluminum Magnesium gauze, Alloy 5056
Name Note 20 mesh woven from 0.23mm (0.009 in.) dia. wire
Avg. Mol. Wt. or Mol. Wt. Range Al:Mg; 95:5 wt%
Quantity 150 x 150 mm
Mesh Size 20 Mesh
Odor Odorless
Form Wire Cloth
MDL Number MFCD00214039