missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Aluminum Keyed Different Safety Lock with Steel Shackle
Help ensure workers stay safe from hazardous energy during servicing and repair with strong, efficient and cost-effective Safety Lockout Padlocks from Brady.
Supplier: Brady™ SDALGRN38STKD
Description
- Designed exclusively for lockout tagout applications as part of an OSHA-compliant safety program.
- Key-retaining brass cylinder prevents key removal in the unlocked position for added safety and helps limit lost keys.
- Aluminum padlocks with steel shackles are durable, corrosion-resistant and ideal for general lockout applications.
- Locks that are Keyed Different (KD) are ideal for simple lockout applications and a manageable number of energy isolation points.
- Locks that are Keyed Alike (KA) reduce the number of keys needed to carry and are ideal for individuals or trades responsible for multiple machines or isolation points.
- Record and retain engraved key codes to streamline replacement of lost keys.
Chemical Identifiers
| Padlock | |
| Aluminum | |
| 6.20 mm | |
| 38.10 mm | |
| 6 |
| Keyed Different | |
| 0.244 in. | |
| 0.75 x 1.5 x 1.8 in. | |
| Steel |
Specifications
| Padlock | |
| Green | |
| 0.244 in. | |
| 0.75 x 1.5 x 1.8 in. | |
| (1) Padlock, (2) Keys | |
| 6 |
| Keyed Different | |
| Aluminum | |
| 6.20 mm | |
| 38.10 mm | |
| Steel | |
| 1 Ea. |