missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Isopropoxide 98.0+%, TCI America™
Supplier: TCI America A0246500G
Specifications
| Aluminum Isopropoxide | |
| White | |
| Crystals | |
| C9H21AlO3 | |
| 1325 | |
| SMZOGRDCAXLAAR-UHFFFAOYSA-N | |
| aluminum;propan-2-olate | |
| 11143 | |
| ≥98.0% (T) |
| 136°C | |
| 555-31-7 | |
| 500 g | |
| MFCD00008870 | |
| aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate | |
| CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] | |
| 204.246 | |
| 204.25 |
Chemical Identifiers
| 555-31-7 | |
| 204.246 | |
| SMZOGRDCAXLAAR-UHFFFAOYSA-N | |
| 11143 | |
| CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] |
| C9H21AlO3 | |
| MFCD00008870 | |
| aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate | |
| aluminum;propan-2-olate |
Safety and Handling
EINECSNumber : (2)-2224&(2)-2388
RTECSNumber : BD0975000
TSCA : Yes