missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Isopropoxide 98.0+%, TCI America™

Catalog No. A0246100G
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0246100G 100 g
A0246500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0246100G Supplier TCI America Supplier No. A0246100G
Only null left

Chemical Identifiers

CAS 555-31-7
Molecular Formula C9H21AlO3
Molecular Weight (g/mol) 204.246
MDL Number MFCD00008870
InChI Key SMZOGRDCAXLAAR-UHFFFAOYSA-N
Synonym aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate
PubChem CID 11143
IUPAC Name aluminum;propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3]

Specifications

Melting Point 136°C
Color White
Physical Form Crystals
Quantity 100 g
UN Number 1325
Formula Weight 204.25
Percent Purity ≥98.0% (T)
Chemical Name or Material Aluminum Isopropoxide
EINECSNumber (2)-2224&(2)-2388
RTECSNumber BD0975000
TSCA Yes