missing translation for 'onlineSavingsMsg'
Learn More

Aluminum(III) Chloride 98.0+%, TCI America™

Catalog No. A1831500G
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1831500G 500 g
A1831100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1831500G Supplier TCI America Supplier No. A1831500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Color White-Yellow
Physical Form Crystalline Powder
Quantity 500 g
UN Number 1726
Formula Weight 133.33
Percent Purity ≥98.0% (T)
Chemical Name or Material Aluminum(III) Chloride
EINECSNumber (1)-0012
RTECSNumber BD0525000
TSCA Yes