missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Aluminum Foil Tape
Description
- Thermally conductive tape helps dissipate heat and improves heating and cooling efficiency to protect temperature sensitive materials
- Resistant to chemicals to protect surfaces during de-paint process and other chemical masking operations
- Highly engineered acrylic adhesive provides long term durability, but removes cleanly after harsh masking applications
- Comprised of a dead soft aluminum foil backing and coated with a highly engineered pressure sensitive acrylic adhesive
- Ideal for heat shielding, heat reflecting, chemical masking, light enhancement, chemical milling, seaming, sealing and de-paint operations.
- Withstands exposure to flame, weathering, moisture and UV
- Heat and light reflective tape protects surfaces and enhances lighting efficiency
- Dead soft aluminum foil backing conforms to curved and uneven surfaces
- Recognized by UL to UL 746C and Classified by UL to UL 723, Class L for low flammability rating
- Contains plastic core center
Specifications
Specifications
| Product Type | Aluminum Foil Tape |
| For Use With (Application) | Painting, Stripping |
| Adhesive Type | Acrylic |
| Color | Silver |
| Dimensions (L x W) | 60 yd. x 2 in. |
| Material | Acrylic |
| Model | 425 |
| Quantity | 24 Rolls/Cs. |
| Thickness (English) | 4.6 mil |