missing translation for 'onlineSavingsMsg'
Learn More

Aluminum foil, 2.0mm (0.08in) thick, Puratronic™, 99.998% (metals basis)

Catalog No. AA11372FI
Change view
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11372FI 50 x 50 mm
AA11372GH 100 x 100 mm
AA11372HP 100 x 500 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA11372FI Supplier Thermo Scientific Chemicals Supplier No. 011372FI

CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifications

Quantity 50 x 50 mm
Odor Odorless
Weight ≈13.5g/50x50mm
Form Foil
IUPAC Name aluminum
Percent Purity 99.998%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Aluminum foil, 2.0mm (0.08 in.) thick, Temper: as rolled
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only