missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum foil, 1.0mm (0.04in) thick, Puratronic™, 99.998% (metals basis)
Form: Approx. 6.75g/50 x 50mm; Temper: as rolled | CAS: 7429-90-5 | Al | 26.98 g/mol
$173.62 - $1237.01
Chemical Identifiers
| CAS | 7429-90-5 |
|---|---|
| Molecular Formula | Al |
| Molecular Weight (g/mol) | 26.98 |
| MDL Number | MFCD00134029 |
| InChI Key | XAGFODPZIPBFFR-UHFFFAOYSA-N |
| Synonym | aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum |
| PubChem CID | 5359268 |
| ChEBI | CHEBI:33629 |
| SMILES | [Al] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA11373FI
|
Thermo Scientific Chemicals
011373FI |
50 x 50 mm |
Each for $173.62
|
|
|||||
|
AA11373GH
|
Thermo Scientific Chemicals
011373GH |
100 x 100 mm |
Each for $383.90
|
|
|||||
|
AA11373HP
|
Thermo Scientific Chemicals
011373HP |
100 x 500 mm |
Each for $1,237.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7429-90-5 | |
| 26.98 | |
| XAGFODPZIPBFFR-UHFFFAOYSA-N | |
| 5359268 | |
| [Al] |
| Al | |
| MFCD00134029 | |
| aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
| CHEBI:33629 |
Specifications
| Silver | |
| Silver-colored foil | |
| Odorless | |
| Foil | |
| MFCD00134029 | |
| XAGFODPZIPBFFR-UHFFFAOYSA-N | |
| aluminium | |
| 5359268 | |
| Aluminum |
| 7429-90-5 | |
| 50 x 50 mm | |
| 99.998% (Metals basis) | |
| Al | |
| aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
| [Al] | |
| 26.98 | |
| CHEBI:33629 |
Safety and Handling
EINECSNumber : 231-072-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only