missing translation for 'onlineSavingsMsg'
Learn More

Aluminum foil, 0.5mm (0.02in) thick, 99.9% (metals basis)

Catalog No. p-4824581
Change view
Click to view available options
Quantity:
50 x 100 mm
50 x 500 mm
Catalog No. Quantity
AA39733FY 50 x 100 mm
AA39733GD 50 x 500 mm
2 options

Catalog No. AA39733FY

Supplier: Thermo Scientific Chemicals 039733FY

Only null left

Form: Approx. 6.7g/50 x 100mm; Temper: as rolled | CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifications

Color Silver
Appearance Silver-colored foil
Quantity 50 x 100 mm
Odor Odorless
Form Foil
Assay Percent Range 99.9% (Metals basis)
IUPAC Name aluminium
Chemical Name or Material Aluminum
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only