missing translation for 'onlineSavingsMsg'
Learn More

Aluminum foil, 0.25mm (0.01in) thick, annealed, 99.99% (metals basis)

Catalog No. p-4408681
Change view
Click to view available options
Quantity:
100 x 100 mm
100 x 500 mm
50 x 50 mm
Catalog No. Quantity
AA40761FI 50 x 50 mm
AA40761GH 100 x 100 mm
AA40761HP 100 x 500 mm
3 options

Catalog No. AA40761FI

Supplier: Thermo Scientific Chemicals 040761FI

Only null left

Form: approx. 1.7g/50x50mm; Temper: soft | CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifications

Color Silver
Appearance Silver-colored foil
Quantity 50 x 50 mm
Odor Odorless
Form Foil
Assay Percent Range 99.99% (Metals basis)
IUPAC Name aluminium
Chemical Name or Material Aluminum, Annealed
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only