missing translation for 'onlineSavingsMsg'
Learn More

Aluminum foil, 0.025mm (0.001in) thick, 30x30cm (12x12in), 99.45% (metals basis)

Catalog No. AA10558KM
Change view
Click to view available options
Quantity:
100 Pc.
25 Pc.
5 Pc.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10558KM 25 Pc.
AA10558KJ 5 Pc.
AA10558KN 100 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA10558KM Supplier Thermo Scientific Chemicals Supplier No. 010558KM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: Approx. 6.17g/30 x 30cm | CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifications

Color Silver
Appearance Silver-colored foil
Quantity 25 Pc.
Odor Odorless
Form Foil
Assay Percent Range 99.45% (Metals basis)
IUPAC Name aluminum
Chemical Name or Material Aluminum
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only