Learn More
Aluminum fluoride trihydrate, 97%
CAS: 15098-87-0 | AlF3H6O3 | 138.022 g/mol
Supplier: Thermo Scientific Chemicals 01149636
Description
Aluminum fluoride trihydrate is used as an additive for the production of aluminum by electrolysis. It is also used to inhibit fermentation and in the preparation of low index films. It serves as a flux ingredient for the removal of magnesium in the refining of aluminum scrap. Further, it is used in the manufacture of aluminum silicates, ceramics and glass. In addition, it acts as a catalyst for organic synthesis and filler in glass industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Aluminum fluoride trihydrate | |
| 15098-87-0 | |
| Powder | |
| AlF3H6O3 | |
| 14,339 | |
| Slightly soluble in water,acids and alkalis. Insoluble in alcohol and acetone. | |
| O.O.O.[F-].[F-].[F-].[Al+3] | |
| 138.022 | |
| 138.02 (83.98 Anhydrous) | |
| Odorless |
| 250°C (in -3H2O) | |
| White | |
| 500 g | |
| MFCD00003426 | |
| aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride | |
| BKLZIAYVINRQEJ-UHFFFAOYSA-K | |
| aluminum;trifluoride;trihydrate | |
| 22002992 | |
| 97% | |
| 2.17 g/mL |
Chemical Identifiers
| 15098-87-0 | |
| 138.022 | |
| BKLZIAYVINRQEJ-UHFFFAOYSA-K | |
| 22002992 | |
| O.O.O.[F-].[F-].[F-].[Al+3] |
| AlF3H6O3 | |
| MFCD00003426 | |
| aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride | |
| aluminum;trifluoride;trihydrate |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H320-H335
EINECSNumber : 232-051-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only