missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Fluoride, Powder, 60%, Spectrum™ Chemical

Catalog No. p-7164961
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg
Packaging:
Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608511 125 g Poly Bottle
18608512 500 g Poly Bottle
18608514 2.5 kg Poly Pail
18608513 50 kg Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608511 Supplier Spectrum Chemical Mfg Cor Supplier No. A1090125GM
Only null left

Chemical Identifiers

CAS 15098-87-0

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 60%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.