missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Chloride, puriss. p.a., Honeywell Fluka™
Puriss. p.a., anhydrous, crystallized,≥99.0% (AT)
$352.90 - $352.90
Chemical Identifiers
| CAS | 7446-70-0 |
|---|---|
| Molecular Formula | AlCl3 |
| Molecular Weight (g/mol) | 133.33 |
| MDL Number | MFCD00003422 |
| InChI Key | VSCWAEJMTAWNJL-UHFFFAOYSA-K |
| Synonym | aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum |
| IUPAC Name | aluminum(3+) trichloride |
| SMILES | [Al+3].[Cl-].[Cl-].[Cl-] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7446-70-0 | |
| 133.33 | |
| VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
| aluminum(3+) trichloride |
| AlCl3 | |
| MFCD00003422 | |
| aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum | |
| [Al+3].[Cl-].[Cl-].[Cl-] |
Specifications
| 7446-70-0 | |
| 1 kg | |
| AlCl3 | |
| 1726 | |
| VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
| aluminum(3+) trichloride | |
| 133.34g/mol | |
| puriss. p.a. | |
| 1mmHg (100°C) |
| Anhydrous Crystallized | |
| AlCl3 | |
| MFCD00003422 | |
| aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum | |
| [Al+3].[Cl-].[Cl-].[Cl-] | |
| 133.33 | |
| ≥99% (AT) | |
| Glass Bottle | |
| Aluminum Chloride |
Safety and Handling
P280-P305 + P351 + P338-P310
H314-H372-H373
EINECSNumber : 231-208-1
RTECSNumber : BD0525000