missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride, Hexahydrate, USP, 95-102%, Spectrum™ Chemical

Catalog No. p-7157004
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600469 100 g Poly Bottle
18600470 500 g Poly Bottle
18600471 2.5 kg Poly Pail
18600472 12 kg Poly Pail
18600474 25 kg Fiber Drum
18600473 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18600469 Supplier Spectrum Chemical Mfg Cor Supplier No. AL225100GM
Only null left

AL225, 7784-13-6, AlCl3·6H2O

Spectrum™ Chemical Aluminum Chloride, Hexahydrate, USP is mainly used in topical formulations to treat hyperhidrosis (excessive sweating). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
IUPAC Name aluminium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

CAS Max % 100%
Quantity 100 g
Percent Purity 95 to 102%
Grade USP
Packaging Poly Bottle