missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Chloride Hexahydrate, Reagent Grade, Ricca Chemical
Specifications
Specifications
| CAS Min % | 98.0 |
| Physical Form | Fine Crystals or Crystalline Powder |
| CAS Max % | 100.0 |
| Quantity | 500 g |
| Packaging | White Poly Wide Mouth Bottle |
| Grade | Reagent |
| Chemical Name or Material | Aluminum Chloride Hexahydrate |
Safety and Handling
| ShelfLife | 60 months |