missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride, Hexahydrate, Reagent Grade, LabChem™

Catalog No. LC108101
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC108101 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC108101 Supplier LabChem Supplier No. LC108101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent grade aluminum chloride hexahydrate for laboratory applications.

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
IUPAC Name aluminium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Boiling Point 182°C
Melting Point 100°C
Color White
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula AlCl3·6H2O
UN Number UN1726
Solubility Information Soluble in water
Formula Weight 241.43
Grade Reagent
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 2.398
Decomposition Information Fume; Hydrogen chloride
Density 2.398g/cm3
Chemical Name or Material Aluminum Chloride, Hexahydrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Avoid breathing dust, fume.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
Store in a well-ventilated place.
Keep container tightly closed.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature