Learn More
Aluminum chloride hexahydrate, Puratronic™, 99.9995% (metals basis)
Description
Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100°C (decomposition) |
| Color | White |
| pH | 2.5 |
| Physical Form | Crystalline Aggregates |
| Quantity | 5 g |
| UN Number | UN3260 |
| Merck Index | 14,337 |
| Solubility Information | Highly soluble in water. Soluble in alcohol,ether,glycerol,propylene glycol and acetonitrile. |
| Formula Weight | 241.43 (133.34 Anhydrous) |
| Percent Purity | 99.9995% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
| EINECSNumber | 231-208-1 |
| RTECSNumber | BD0530000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only