Learn More
Aluminum chloride hexahydrate, 99%
CAS: 7784-13-6 | AlCl3H12O6 | 241.42 g/mol
Supplier: Thermo Scientific Chemicals A1443730
Description
Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.
Solubility
Highly soluble in water. Soluble in alcohol, ether, glycerol, propylene glycol and acetonitrile.
Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
Specifications
| Aluminum chloride hexahydrate | |
| 7784-13-6 | |
| 250 g | |
| MFCD00149134 | |
| 14,337 | |
| JGDITNMASUZKPW-UHFFFAOYSA-K | |
| aluminium(3+) hexahydrate trichloride | |
| 241.43 (133.34 Anhydrous) | |
| Odorless |
| 100°C (decomposition) | |
| 2.5 | |
| AlCl3H12O6 | |
| UN3260 | |
| Highly soluble in water. Soluble in alcohol,ether,glycerol,propylene glycol and acetonitrile. | |
| O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] | |
| 241.42 | |
| 99% | |
| 2.398 |
Chemical Identifiers
| 7784-13-6 | |
| 241.42 | |
| JGDITNMASUZKPW-UHFFFAOYSA-K |
| AlCl3H12O6 | |
| MFCD00149134 | |
| O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 231-208-1
RTECSNumber : BD0530000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only