missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride Hexahydrate, 99%, Reagent Grade, Honeywell™

Catalog No. p-7120138
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045448 100 g Poly Bottle
60045449 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045448 Supplier Honeywell Chemicals Supplier No. 237078100G
Only null left

Reagent Grade, 99%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Color White
pH 2.5 to 3.5 at 20°C
Quantity 100 g
UN Number NONH for all modes of transport
IUPAC Name aluminum(3+) hexahydrate trichloride
Formula Weight 241.43g/mol
Percent Purity 99%
Grade ReagentPlus
Packaging Poly Bottle
Appearance Crystals or Crystalline Chunk(s) or Chunk(s) or Solid
Vapor Pressure 1mmHg (100°C)
Density 2.4g/cm3
Chemical Name or Material Aluminum Chloride Hexahydrate
Show More Show Less
Hazard Statement P305 + P351 + P338
Precautionary Statement H315-H319
EINECSNumber 231-208-1