missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride, Hexahydrate, 95-102%, Spectrum™ Chemical

Catalog No. p-7165079
Change view
Click to view available options
Quantity:
25 kg
50 kg
Packaging:
Fiber Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608507 25 kg Fiber Drum
18608508 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18608507 Supplier Spectrum Chemical Mfg Cor Supplier No. A402625KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7784-13-6

Specifications

CAS Max % 1
Quantity 25 kg
Percent Purity 95 to 102%
Grade Reagent
Identification To Pass Test
Packaging Fiber Drum

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.