missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride, anhydrous, granular, 99%

Catalog No. p-7081762
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1189230 250 g
AAA118920B 1000 g
AAA118920I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1189230 Supplier Thermo Scientific Chemicals Supplier No. A1189230

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7446-70-0 | AlCl3 | 133.33 g/mol

Aluminum chloride is used as a catalyst for Friedel-Crafts acylation and alkylation of aromatic compounds. It is one of the most commonly employed Lewis acids for a wide variety of organic transformations. It catalyzes the ene reaction, polymerization and isomerization reactions. For example, it can be used for the synthesis of ethyl benzene which is a precursor for producing polystyrene. It can be used for producing dodecylbenzene, a key intermediate for detergents. It is useful in the production of anthroquinone, the presursor for dyestuffs. It is used in the synthesis of bis (arene) metal complexes, through Fischer-Hafner synthesis. Gattermann-Koch reaction employs aluminum chloride for introducing formyl group onto aromatic rings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aluminum chloride is used as a catalyst for Friedel-Crafts acylation and alkylation of aromatic compounds. It is one of the most commonly employed Lewis acids for a wide variety of organic transformations. It catalyzes the ene reaction, polymerization and isomerization reactions. For example, it can be used for the synthesis of ethyl benzene which is a precursor for producing polystyrene. It can be used for producing dodecylbenzene, a key intermediate for detergents. It is useful in the production of anthroquinone, the presursor for dyestuffs. It is used in the synthesis of bis (arene) metal complexes, through Fischer-Hafner synthesis. Gattermann-Koch reaction employs aluminum chloride for introducing formyl group onto aromatic rings.

Solubility
Soluble in water, hydrochloric acid and ethanol. Slightly soluble in benzene.

Notes
Moisture sensitive. Reacts violently with water with liberation of heat.

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Melting Point 190°C (sublimation)
pH 2.4
Physical Form Granules
Quantity 250 g
Odor Pungent
Sensitivity Moisture sensitive
Density 2.44
UN Number UN1726
Merck Index 14,337
Solubility Information Soluble in water,hydrochloric acid and ethanol. Slightly soluble in benzene.
Formula Weight 133.34
Percent Purity 99%
Chemical Name or Material Aluminum chloride, Anhydrous
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ALUMINUM CHLORIDE, ANHYDROUS
EINECSNumber 231-208-1
RTECSNumber BD0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only