missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride, Anhydrous, 95%, Spectrum™ Chemical

Catalog No. 18608501
Change view
Click to view available options
Quantity:
100 g
500 g
45 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608501 500 g Amber Glass Bottle
18608500 100 g Amber Glass Bottle
18608499 45 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608501 Supplier Spectrum Chemical Mfg Cor Supplier No. A1078500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7446-70-0

Specifications

CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 500 g
Molecular Weight (g/mol) 133.34
Percent Purity 95%
Grade Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Aluminum Chloride, Anhydrous

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.