Learn More
Aluminum chloride, 98+%, extra pure, anhydrous powder
Description
Aluminum chloride is used to control excessive sweating. It is commonly used as a catalyst for the halogenation (especially chlorination) of aromatic groups, as well as in the Friedel Crafts reaction. It is also used in the Meerwein-Ponndorf-Verley reduction. It is widely used in the manufacturing of petrochemicals such as alkylbenzene, ethylbenzene, alkyl aryl ketone, ethyl chloride. It is used for making detergents, polymerization and isomerization reactions of hydrocarbons.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aluminum chloride is used to control excessive sweating. It is commonly used as a catalyst for the halogenation (especially chlorination) of aromatic groups, as well as in the Friedel Crafts reaction. It is also used in the Meerwein-Ponndorf-Verley reduction. It is widely used in the manufacturing of petrochemicals such as alkylbenzene, ethylbenzene, alkyl aryl ketone, ethyl chloride. It is used for making detergents, polymerization and isomerization reactions of hydrocarbons.
Solubility
Soluble in hydrochloric acid, ethanol, chloroform and carbon tetrachloride.Slightly soluble in benzene.
Notes
Aluminum chloride is hygroscopic. It must be packed in a tightly closed container and stored in a cool, dry, and well-ventilated area. It is water-reactive and corrosive in nature. It reacts vigorously with bases.
Specifications
Specifications
| Melting Point | 190°C (sublimation) |
| pH | 2.4 |
| Physical Form | Powder |
| Quantity | 500 g |
| Odor | Pungent |
| Sensitivity | Moisture sensitive |
| Density | 2.44 |
| UN Number | UN1726 |
| Merck Index | 14,337 |
| Solubility Information | Soluble in hydrochloric acid,ethanol,chloroform and carbon tetrachloride.Slightly soluble in benzene. |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314 Causes severe skin burns and eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ALUMINUM CHLORIDE, ANHYDROUS |
| EINECSNumber | 231-208-1 |
| RTECSNumber | BD0525000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only