missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride, 0.5M aq. soln., Thermo Scientific™

Catalog No. AAJ61582AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61582AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61582AE Supplier Thermo Scientific Chemicals Supplier No. J61582AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Color Colorless
Physical Form Liquid
Quantity 100 mL
UN Number UN2581
IUPAC Name aluminium(3+) hexahydrate trichloride
Formula Weight 133.34
Concentration or Composition (by Analyte or Components) 0.5M aq. soln.
Chemical Name or Material Aluminum chloride
Hazard Category H315-H319
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALUMINUM CHLORIDE, SOLUTION
TSCA Yes
Recommended Storage Ambient temperatures