missing translation for 'onlineSavingsMsg'
Learn More

Aluminum carbide, 99+% (metals basis)

Catalog No. p-7049336
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1403818 50 g
AA1403830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1403818 Supplier Thermo Scientific Chemicals Supplier No. 01403818

CAS: 1299-86-1 | C3Al4 | 143.96 g/mol

Aluminum carbide is used as an abrasive material for cutting tools. It is also used to reduce metal oxides and to prepare aluminum nitride and methane. It and also finds use in metallurgy. Further, it acts as a catalyst and a drying agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1299-86-1
Molecular Formula C3Al4
Molecular Weight (g/mol) 143.96
MDL Number MFCD00003425
InChI Key CAVCGVPGBKGDTG-UHFFFAOYSA-N
Synonym aluminum carbide, 1,3,5,7-tetraaluminahepta-3,4-dien-1,6-diyne, aluminum carbide,al4c3, al4c3 aluminum carbide, aluminum carbide, al4c3, aluminum carbide, powder, ∼325 mesh
PubChem CID 16685054
IUPAC Name alumanylidynemethyl(alumanylidynemethylalumanylidenemethylidene)alumane
SMILES [Al]#C[Al]=C=[Al]C#[Al]

Specifications

Melting Point 2100°C (>2200° (decomposition))
Color Gray
Quantity 50 g
UN Number UN1394
Merck Index 14,335
Solubility Information Soluble in dilute hydrochloric acid.
Formula Weight 143.96
Percent Purity ≥99%
Sensitivity Moisture sensitive
Density 2.36 g/mL
Assay (metals basis)
Chemical Name or Material Aluminum carbide
Show More Show Less
Hazard Category H228-H261-H315-H319-H335
Hazard Statement GHS H Statement
H228-H261-H315-H319-H335
Flammable solid.
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P223-P231+P232-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P335+P334-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ALUMINUM CARBIDE
EINECSNumber 215-076-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only