missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Bromide Anhydrous 95.0+%, TCI America™

Catalog No. p-7118026
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26925G 5 g
A269225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26925G Supplier TCI America Supplier No. A26925G
Only null left

Chemical Identifiers

CAS 7727-15-3
Molecular Formula AlBr3
Molecular Weight (g/mol) 266.69
MDL Number MFCD00003421
InChI Key PQLAYKMGZDUDLQ-UHFFFAOYSA-K
IUPAC Name aluminium(3+) tribromide
SMILES [Al+3].[Br-].[Br-].[Br-]

Specifications

Boiling Point 256°C
Melting Point 98°C
Physical Form Crystalline Powder
Quantity 5 g
UN Number 1725
Formula Weight 266.69
Percent Purity ≥95.0% (T)
Chemical Name or Material Aluminum Bromide Anhydrous
EINECSNumber (1)-1022
RTECSNumber BD0350000
TSCA Yes