missing translation for 'onlineSavingsMsg'
Learn More

Aluminum bromide, 99.997% (metals basis)

Catalog No. aa8906514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8906514 25 g
AA8906506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8906514 Supplier Thermo Scientific Chemicals Supplier No. 08906514
Only null left

CAS: 7727-15-3 | AlBr3 | 266.69 g/mol

Aluminum Bromide, is used as a catalyst for the Friedel-Crafts alkylation reaction, and also for bromination and isomerization in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7727-15-3
Molecular Formula AlBr3
Molecular Weight (g/mol) 266.69
MDL Number MFCD00003421
InChI Key PQLAYKMGZDUDLQ-UHFFFAOYSA-K
Synonym Aluminum tribromide
IUPAC Name aluminium(3+) tribromide
SMILES [Al+3].[Br-].[Br-].[Br-]

Specifications

Boiling Point 265°C
Melting Point 97.5°C
Color Orange to White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.997% (Metals basis)
Solubility Information Soluble in benzene,alcohol,carbon disulfide,ether and toluene. Reacts with water.
Formula Weight 266.71
Odor Pungent
Sensitivity Moisture sensitive
Density 3.21 g/cm3
Chemical Name or Material Aluminum bromide
Show More Show Less
Hazard Category H302-H314
Hazard Statement Keep container dry. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 231-779-7
RTECSNumber BD0350000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only