missing translation for 'onlineSavingsMsg'
Learn More

Aluminum bromide, 98%

CAS: 7727-15-3 | AlBr3 | 266.69 g/mol

Supplier:  Thermo Scientific Chemicals 01111309

 View more versions of this product

Catalog No. AA1111309


Only null left
Explore available promotions

Description

Description

Alkylation, bromination and isomerization catalyst in organic synthesis.Aluminum bromide acts as a strong Lewis acid and is used in Lewis acid-promoted reactions such as epoxide ring openings. It is used as a catalyst for Friedel-Crafts alkylation reaction. It also finds application in bromination and isomerization reactions in organic synthesis. It is also used in polymerization reaction to prepare poly(o-xylene)from benzocyclobutene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

265°C
97.5°C
Orange to White
10 g
AlBr3
Aluminum tribromide
PQLAYKMGZDUDLQ-UHFFFAOYSA-K
aluminium(3+) tribromide
266.71
Moisture sensitive
3.21 g/cm3
Aluminum bromide
7727-15-3
Crystalline
98%
MFCD00003421
Reacts with water violently,soluble in alcohol,carbon disulfide,ether.Slightly soluble in methanol,diethyl ether and acetone.
[Al+3].[Br-].[Br-].[Br-]
266.69
Pungent
Reacts with water violently

Chemical Identifiers

7727-15-3
266.69
PQLAYKMGZDUDLQ-UHFFFAOYSA-K
aluminium(3+) tribromide
AlBr3
MFCD00003421
Aluminum tribromide
[Al+3].[Br-].[Br-].[Br-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Keep container dry. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 231-779-7

RTECSNumber : BD0350000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only