missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Aluminum bromide, 98%

Catalog No. AA1111359
Change view
Click to view available options
Quantity:
5 x 100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1111359 5 x 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1111359 Supplier Thermo Scientific™ Supplier No. 1111359
Alkylation, bromination and isomerization catalyst in organic synthesis

Specifications

Boiling Point 265°
Melting Point 97.5°
CAS 7727-15-3
Physical Form Solid
Quantity 5 x 100 g
Molecular Formula AlBr{3}
MDL Number MFCD00003421
UN Number UN1725
Fieser 1,22 2,19 3,7 4,10 5,10 6,17 9,11
Merck Index 14,332
Synonym Aluminum tribromide;
Formula Weight 266.71
Percent Purity 98%
Density 3.21
Show More Show Less