Learn More
Aluminum antimonide, 99.99% (metals basis), Thermo Scientific Chemicals
In semiconductor technology
$310.70 - $310.70
Chemical Identifiers
| CAS | 25152-52-7 |
|---|---|
| Molecular Formula | AlSb |
| Molecular Weight (g/mol) | 148.742 |
| MDL Number | MFCD00014154 |
| InChI Key | LVQULNGDVIKLPK-UHFFFAOYSA-N |
| Synonym | aluminum antimonide, aluminum antimonide, unii-5yy752fd6d, aluminum, compd. with antimony 1:1, aluminiumantimonid |
| PubChem CID | 91307 |
| IUPAC Name | alumanylidynestibane |
| SMILES | [Al]#[Sb] |
Description
In semiconductor technology
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 25152-52-7 | |
| 148.742 | |
| LVQULNGDVIKLPK-UHFFFAOYSA-N | |
| 91307 | |
| [Al]#[Sb] |
| AlSb | |
| MFCD00014154 | |
| aluminum antimonide, aluminum antimonide, unii-5yy752fd6d, aluminum, compd. with antimony 1:1, aluminiumantimonid | |
| alumanylidynestibane |
Specifications
| 1050°C | |
| Pieces | |
| AlSb | |
| UN1549 | |
| LVQULNGDVIKLPK-UHFFFAOYSA-N | |
| alumanylidynestibane | |
| 91307 | |
| 99.99% | |
| (metals basis) |
| 25152-52-7 | |
| 5 g | |
| MFCD00014154 | |
| aluminum antimonide, aluminum antimonide, unii-5yy752fd6d, aluminum, compd. with antimony 1:1, aluminiumantimonid | |
| [Al]#[Sb] | |
| 148.742 | |
| 148.73 | |
| Aluminum antimonide |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
EINECSNumber : 246-667-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only