missing translation for 'onlineSavingsMsg'
Learn More

Aluminum antimonide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. AA4345506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4345506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4345506 Supplier Thermo Scientific Chemicals Supplier No. 04345506

In semiconductor technology

In semiconductor technology

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25152-52-7
Molecular Formula AlSb
Molecular Weight (g/mol) 148.742
MDL Number MFCD00014154
InChI Key LVQULNGDVIKLPK-UHFFFAOYSA-N
Synonym aluminum antimonide, unii-5yy752fd6d, aluminum, compd. with antimony 1:1, aluminiumantimonid
PubChem CID 91307
IUPAC Name alumanylidynestibane
SMILES [Al]#[Sb]

Specifications

Melting Point 1050°C
Physical Form Pieces
Quantity 5 g
UN Number UN1549
Formula Weight 148.73
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Aluminum antimonide
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
EINECSNumber 246-667-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only