Learn More
Aluminum acrylate
CAS: 15743-20-1 | C9H12AlO6 | 243.171 g/mol
Supplier: Thermo Scientific Chemicals 04200314
Description
Aluminum acrylate is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Aluminum acrylate | |
| C9H12AlO6 | |
| 25 g | |
| aluminum acrylate | |
| UNGWYUCJMHZVEX-UHFFFAOYSA-N | |
| aluminum;prop-2-enoic acid | |
| 131874419 | |
| Powder |
| 15743-20-1 | |
| MFCD00078421 | |
| Air and moisture sensitive | |
| soluble in acetone,benzene,carbon tetrachloride. Miscible with alcohol,ether,ethanol,ethyl ether,chloroform. | |
| C=CC(=O)O.C=CC(=O)O.C=CC(=O)O.[Al] | |
| 243.171 | |
| 243.17 |
Chemical Identifiers
| 15743-20-1 | |
| 243.171 | |
| UNGWYUCJMHZVEX-UHFFFAOYSA-N | |
| 131874419 | |
| C=CC(=O)O.C=CC(=O)O.C=CC(=O)O.[Al] |
| C9H12AlO6 | |
| MFCD00078421 | |
| aluminum acrylate | |
| aluminum;prop-2-enoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 239-836-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only