missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Acetate, Dibasic, Stabilized with Boric Acid, Powder, Approx. 30%, Spectrum™ Chemical

Catalog No. 18600460
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
150 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600460 125 g Amber Glass Bottle
18600461 500 g Amber Glass Bottle
18600464 150 lb. Fiber Drum
18600462 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600460 Supplier Spectrum Chemical Mfg Cor Supplier No. AL223125GM
Only null left

AL223, C2H5O4Al·H3BO3

Spectrum™ Chemical Aluminum Acetate, Dibasic, Stabilized with Boric Acid, Powder is a salt that is used as a topical astringent to treat skin irritations from poison ivy, substances such as soaps and cosmetics and insect bites. Aluminum acetate is particularly helpful for foot problems including athlete’s foot, swelling, and excessive sweating. It is also used to treat ear canal infections. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 7360-44-3 , 10043-35-3

Specifications

CAS Max % 30%, 70%
Quantity 125 g
Percent Purity ∼30%
Grade Reagent
Packaging Amber Glass Bottle