Learn More
Aluminum tri-sec-butoxide, 97%
CAS: 2269-22-9 | C12H27AlO3 | 246.33 g/mol
$181.02 - $181.02
Chemical Identifiers
| CAS | 2269-22-9 |
|---|---|
| Molecular Formula | C12H27AlO3 |
| Molecular Weight (g/mol) | 246.33 |
| MDL Number | MFCD00009327 |
| InChI Key | LWGPRERTOLVRLK-UHFFFAOYSA-N |
| Synonym | aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide |
| PubChem CID | 50931103 |
| IUPAC Name | aluminum;butan-2-ol |
| SMILES | CCC(C)O.CCC(C)O.CCC(C)O.[Al] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC191215000
|
Thermo Scientific Chemicals
191215000 |
500 g | Glass bottle |
Each for $181.02
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
| 2269-22-9 | |
| 246.33 | |
| LWGPRERTOLVRLK-UHFFFAOYSA-N | |
| 50931103 | |
| CCC(C)O.CCC(C)O.CCC(C)O.[Al] |
| C12H27AlO3 | |
| MFCD00009327 | |
| aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide | |
| aluminum;butan-2-ol |
Specifications
| 2269-22-9 | |
| 0.9600g/mL | |
| 27°C | |
| 96% min. (ex Al) | |
| C12H27AlO3 | |
| [C2H5CH(CH3)O]3Al | |
| 500 g | |
| 0.96 | |
| Solubility in water: decomposes | |
| CCC(C)O.CCC(C)O.CCC(C)O.[Al] | |
| 246.33 | |
| 1800 mPa.s (20°C) | |
| 97% | |
| Aluminum tri-sec-butoxide |
| Yellow | |
| 200.0°C to 206.0°C (30.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4380 to 1.4400 | |
| MFCD00009327 | |
| 01,IV,1567 | |
| aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide | |
| LWGPRERTOLVRLK-UHFFFAOYSA-N | |
| aluminum;butan-2-ol | |
| 50931103 | |
| 246.33 | |
| Oily Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
If eye
GHS Signal Word: Warning
EINECSNumber : 218-871-2
RUO – Research Use Only