missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A

Catalog No. p-4258664
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC365750010 1 kg Plastic bottle
AC365750025 2.5 kg Plastic bottle
AC365750100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC365750010 Supplier Thermo Scientific Chemicals Supplier No. 365750010
Only null left

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for column chromatography

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980.0°C
Melting Point 2030.0°C
Color White
pH 4.0 to 5.0 (5% aq. suspension, stirred)
Physical Form Powder
Quantity 1 kg
Linear Formula Al2O3
Fieser 01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9
Merck Index 15,35
Solubility Information Solubility in water: insoluble. Other solubilities: slightly sol. in alkaline sol.(forming hydroxides), practically insoluble in non-polar organic solvent, difficultly soluble in mineral acids
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Grade Chromatography
Packaging Plastic bottle
Chemical Name or Material Aluminum oxide
Show More Show Less
EINECSNumber 215-691-6

RUO – Research Use Only