missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum oxide, 99.99%, (trace metal basis), extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Boiling Point | 2977.0°C |
| Melting Point | 2030.0°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.99% |
| Linear Formula | AI2O3 |
| Fieser | 01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9 |
| Merck Index | 15, 350 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: slightly sol. in alkaline sol.(forming hydroxides), practically insoluble in non-polar organic, solvents, difficultly soluble in mineral acids |
| Show More |
Safety and Handling
| EINECSNumber | 215-691-6 |
RUO – Research Use Only