missing translation for 'onlineSavingsMsg'
Learn More

Aluminum foil, For analysis, 0.3mm thickness, 30mm width, EMSURE™, MilliporeSigma™

Catalog No. M1010570250
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bag
Plastic box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1010570250 250 g Plastic box
M1010571000 1 kg Plastic bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1010570250 Supplier Technidata Supplier No. 1.01057.0250
For analysis

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminium
SMILES [Al]

Specifications

Appearance Silver solid
Quantity 250 g
Odor Odorless
Packaging Plastic box
Form Solid
Formula Weight 26.98
Chemical Name or Material Aluminum