missing translation for 'onlineSavingsMsg'
Learn More

Aluminum, Fine Powder, Stabilized About 2% fat, MilliporeSigma™

Catalog No. M1010561000
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Metal can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1010561000 1 kg Metal can
M1010560250 250 g Metal can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1010561000 Supplier Technidata Supplier No. 1.01056.1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminium
SMILES [Al]

Specifications

Appearance Metallic powder
Quantity 1 kg
Odor Odorless
Packaging Metal can
Form Solid
UN Number UN 1396
Formula Weight 26.98
Chemical Name or Material Aluminum
Hazard Category H228: Flammable solid.
H261: In contact with water releases flammable gases.
DOTInformation DOT Class: 4.3, Packaging Group: II
EINECSNumber 231-072-3