Learn More
Aluminum chloride, 98.5%, extra pure, anhydrous, powder
CAS: 7446-70-0 | AlCl3 | 133.33 g/mol
Supplier: Thermo Scientific Chemicals 217461000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Lewis acidSpecifications
| Aluminum chloride | |
| 7446-70-0 | |
| Powder | |
| 98% min. (Argentometry) | |
| AlCl3 | |
| 01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 07,7; 08,13; 09,11; 10,9; 11,25; 12,26; 13,15; 14,20; 15,10; 16,10; 17,15 | |
| aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum | |
| VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
| aluminum(3+) trichloride | |
| 133.34 | |
| Extra Pure | |
| 2.44 |
| 194.0°C | |
| Gray or White to Yellow | |
| 100 g | |
| AlCl3 | |
| MFCD00003422 | |
| 15, 333 | |
| Solubility in water: reacts. Other solubilities: freely miscible with benzene, nitrobenzene,, benzophenone, carbon tetrachloride and chloroform | |
| [Al+3].[Cl-].[Cl-].[Cl-] | |
| 133.33 | |
| 98.5% | |
| Glass bottle | |
| 2.4400g/mL |
Chemical Identifiers
| 7446-70-0 | |
| 133.33 | |
| VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
| aluminum(3+) trichloride |
| AlCl3 | |
| MFCD00003422 | |
| aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum | |
| [Al+3].[Cl-].[Cl-].[Cl-] |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 231-208-1
RTECSNumber : BD0525000
TSCA : TSCA
RUO – Research Use Only