missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride, 98.5%, extra pure, anhydrous, powder

Catalog No. AC217460050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC217460050 5 g Glass bottle
AC217461000 100 g Glass bottle
AC217465000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC217460050 Supplier Thermo Scientific Chemicals Supplier No. 217460050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7446-70-0 | AlCl3 | 133.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Lewis acid

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Melting Point 194.0°C
Color Gray or White to Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 98% min. (Argentometry)
Linear Formula AlCl3
Fieser 01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 07,7; 08,13; 09,11; 10,9; 11,25; 12,26; 13,15; 14,20; 15,10; 16,10; 17,15
Merck Index 15, 333
Solubility Information Solubility in water: reacts. Other solubilities: freely miscible with benzene, nitrobenzene,, benzophenone, carbon tetrachloride and chloroform
Formula Weight 133.34
Percent Purity 98.5%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 2.44
Density 2.4400g/mL
Chemical Name or Material Aluminum chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 231-208-1
RTECSNumber BD0525000
TSCA TSCA

RUO – Research Use Only