Learn More
Solstice Alumina SPE Columns, Alumina Neutral, 1000 mg/6 mL
Catalog No. 6002711
Change view
Click to view available options
Quantity:
30
Catalog No. Quantity
6002711 30
1 options
Catalog No. 6002711 Supplier Solstice Supplier No. 9138
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Medical-grade polypropylene with PE frits
  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality
Specifications
Quantity 30