missing translation for 'onlineSavingsMsg'
Learn More

Alumina N, Activity: Super I, MP EcoChrom™, MP Biomedicals™

Catalog No. ICN04580
Change view
Click to view available options
Quantity:
500 g
5 kg
50 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN04580 500 g
ICN04586 5 kg
ICN04589 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN04580 Supplier MP Biomedicals Inc Supplier No. 0204580
Only null left

MP EcoChrom™ Alumina N, Activity Super I are commonly used for purification processes of all kinds, including sample preparation for pesticides, and environmental analysis, solvent purification and all kinds of chromatography, requiring the highest precision and reproducibility.

  • MP EcoChrom™ Alumina N, Activity Super I shows a sorbtion capacity approximately twice as high as that of MP EcoChrom™ Alumina N, Activity I.
  • Their inital activity is kept in a very narrow range and their dactivation behavior is extremely constant.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Free-flowing Powder
Quantity 500 g
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina B, Activity: Super I
EINECSNumber 215-691-6
Recommended Storage Store at room temperature.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.