missing translation for 'onlineSavingsMsg'
Learn More

Alumina N, Activity: I, MP EcoChrom™, MP Biomedicals™

Catalog No. p-4650840
Change view
Click to view available options
Quantity:
1 kg
5 kg
50 kg
500 g
Catalog No. Quantity
ICN02084 500 g
ICN02087 1 kg
ICN02090 5 kg
ICN02135 50 kg
4 options

Catalog No. ICN02084

Supplier: MP Biomedicals Inc 0202084

Only null left

MP EcoChrom™ Alumina is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc.

  • MP EcoChrom™ Alumina is widely used as an adsorbent, desiccant, abrasive, electrical insulator and as a catalyst for various organic reactions.
  • It is also used for isolation of alkaloids, glycosides, active substances from plants, vitamins, hormones and antibiotics, dehydration of organic solvents, purification of enzymes, removal of terpenes, peroxides, pyrogens from injectable solutions and infusions.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Free-flowing Powder
Quantity 500 g
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina B, Activity: I
EINECSNumber 215-691-6
Recommended Storage Store at room temperature.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.