Learn More
Perkin Elmer Canada ALUMINA INJECTOR,1.2 MM I.D. DFS Item
Supplier:  Perkin Elmer Canada N0781015
Alumina Injector, 1.2 mm i.d.For use with the Optima 8x00 ICP-OES.Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents, e.g., xylene.
Catalog No. NC1453283
Only null left
Explore available promotions