missing translation for 'onlineSavingsMsg'
Learn More

CoorsTek™ Alumina Conical Crucible

High-grade aluminum oxide yields contamination-free results

Supplier:  CoorsTek™ 65518

 View more versions of this product

Catalog No. 12-220H


Only null left

Description

Description

Highly resistant to chemical attack, this alumina-ceramic crucible is especially useful to chemists, metallurgists, and others involved in high-temperature work

  • 99.8% pure aluminum oxide
  • Withstands temperatures up to 1750°C
  • Inert in hydrogen and carbonaceous atmospheres
  • Offers high resistance to alkalies and other fluxes
  • Permeability: gas tight
  • Water absorption: none

Suitable for glass melting, including borosilicate glass. Recommended for use with refractory metals such as molybdenum, platinum, rhodium, tungsten, tantalum and iridium

Specifications

Specifications

Alumina
1750°C
53mm
33mm
For determinations involving molybdenum, platinum, rhodium, tungsten, tantalum and iridium
50 mL
Conical Crucible
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

High-Alumina Labware temperature change rate should not exceed 150°C/hour