missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alumina B, Activity: Super I, Basic pH, MP EcoChrom™, MP Biomedicals™
Description
- MP EcoChrom™ Alumina - Super I is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc.
- MP Alumina B - Super I show absorption capacity approximately twice as high as an Alumin B, Activity I.
- The initial activity is kept in a very narrow range and their deactivation behaviour is extremely constant.
- The product is available as a neutral version, MP Alumina N - Super I, and an acidic version, MP Alumina A - Super I.
- MP EcoChro Alumina B - Super I is commonly used for purification processes of all kinds including sample preparation for pesticides, environmental analysis, solvent purification and all kinds of chromatography requiring the highest precision and reproducibility.
Specifications
Specifications
| Color | Pink to White |
| Physical Form | Free-flowing Powder |
| Quantity | 5 kg |
| IUPAC Name | dialuminum;oxygen(2-) |
| Formula Weight | 101.96 |
| Density | 3.94 g/cm3 (at 20°C) |
| Chemical Name or Material | Alumina B, Activity: Super I |
Safety and Handling
| EINECSNumber | 215-691-6 |
| Recommended Storage | Store at room temperature. |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.