missing translation for 'onlineSavingsMsg'
Learn More

Alumina B, Activity: Super I, Dioxin Analysis, MP EcoChrom™, 63 to 200 μm, MP Biomedicals™

Catalog No. ICN04569
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
ICN04569 500 g
1 options

Catalog No. ICN04569

Supplier: MP Biomedicals Inc 0204569

Only null left

MP EcoChrom™ Alumina is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc. This Alumina is produced at the highest activity available in the market.

  • MP EcoChrom™ Alumina is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc.
  • MP Alumina B - Super I for Dioxin Analysis is an Alumina produced at the highest activity available in the market.
  • This product meets the high recovery level required for the analysis of polychlorinated dibenzodioxins (PCDD) and dibenzofurans (PCDF) according to VDI regulation 3499, part I.
  • MP EcoChrom Alumina B - Super I for Dioxin Analysis is used for the sample preparation of Dioxin Analysis.
  • It is also suitable for all chromatographic processes requiring highest activity and/or highest reproducibility.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Free-flowing Powder
Quantity 500 g
IUPAC Name dialuminium(3+) trioxidandiide
Formula Weight 101.96
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina B, Activity: Super I
EINECSNumber 215-691-6
Recommended Storage Store at room temperature.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.